Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olaparib-d5 | 2143107-56-4 | MFCD34469897; MFCD12912443 | 99.4% | 439.49 g·mol-1 | C24H18D5FN4O3 | 1 MG
SDP

Catalog No. 5000247784
Encompass_Preferred

Olaparib-d5 is the deuterium-labeled analogue of olaparib intended for use as a stable isotope internal standard in quantitative LC-MS/MS and pharmacokinetic studies. It offers a mass-shifted counterpart to the parent drug while maintaining similar chromatographic properties, enabling precise quantitation and method validation in analytical workflows.

  • Stable isotope-labeled internal standard for LC-MS/MS assays.
  • Provides a mass-shifted analogue for accurate quantitation.
  • Near-identical chromatographic behavior to the parent compound.
  • High purity suitable for analytical and pharmacokinetic studies.
  • Soluble in DMSO and DMF; limited solubility in ethanol.
  • Recommended storage: powder at -20°C; in solution at -80°C for extended storage.

Catalog No. 50-002-47784 Supplier Medchemexpress LLC Supplier No. HY10162S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olaparib-d5 is the deuterium-labeled analogue of olaparib intended for use as a stable isotope internal standard in quantitative LC-MS/MS and pharmacokinetic studies. It offers a mass-shifted counterpart to the parent drug while maintaining similar chromatographic properties, enabling precise quantitation and method validation in analytical workflows.

  • Stable isotope-labeled internal standard for LC-MS/MS assays.
  • Provides a mass-shifted analogue for accurate quantitation.
  • Near-identical chromatographic behavior to the parent compound.
  • High purity suitable for analytical and pharmacokinetic studies.
  • Soluble in DMSO and DMF; limited solubility in ethanol.
  • Recommended storage: powder at -20°C; in solution at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.