Learn More
Medchemexpress LLC Olaparib-d5 | 2143107-56-4 | MFCD34469897; MFCD12912443 | 99.4% | 439.49 g/mol | C24H18D5FN4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10162S10MG
Encompass_Preferred
Olaparib-d5 is a deuterium-labeled analogue of the PARP inhibitor olaparib, intended as an isotopically labeled internal standard or tracer for quantitative analytical applications. It is formulated for use in NMR, GC-MS, and LC-MS workflows, and has molecular formula C24H18D5FN4O3, molecular weight 439.49, and CAS 2143107-56-4.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for trace-level detection and quantitation.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • Reduces chromatographic and mass spectral interferences in assays.
  • Provided as a small research quantity for method development and validation.
Catalog No. 50-002-49074