Learn More
Medchemexpress LLC Olaparib-d5 (AZD2281-d5) | 2143107-56-4 | MFCD34469897 | 99.4% | 439.49 | C24H18D5FN4O3 | 5 MG
SDP
Catalog No. 5000246998
Encompass_Preferred
Olaparib-d5 is the deuterium-labeled stable isotope of the PARP inhibitor olaparib, supplied as a high-purity analytical reference standard for method development, quantitative analysis, and use as an internal standard in mass spectrometry workflows. It is provided in small, accurately weighed packages suitable for LC-MS and other sensitive analytical techniques.
  • High purity (99.37%) for reliable analytical results.
  • Deuterium labeling produces a clear mass shift for internal standard use in MS.
  • Suitable for LC-MS method development and quantitative assays.
  • Soluble in DMSO, DMF, and ethanol for flexible sample preparation.
  • Available in small mg packages for precise weighing and low consumption.
Catalog No. 50-002-46998 Supplier Medchemexpress LLC Supplier No. HY10162S5MG
Olaparib-d5 is the deuterium-labeled stable isotope of the PARP inhibitor olaparib, supplied as a high-purity analytical reference standard for method development, quantitative analysis, and use as an internal standard in mass spectrometry workflows. It is provided in small, accurately weighed packages suitable for LC-MS and other sensitive analytical techniques.
  • High purity (99.37%) for reliable analytical results.
  • Deuterium labeling produces a clear mass shift for internal standard use in MS.
  • Suitable for LC-MS method development and quantitative assays.
  • Soluble in DMSO, DMF, and ethanol for flexible sample preparation.
  • Available in small mg packages for precise weighing and low consumption.