Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olaparib-d8 | 00-00-0 | 99.6% | 442.51 g/mol | C24H15D8FN4O3 | 1 MG
SDP

Catalog No. 5000247783
Encompass_Preferred

Olaparib-d8 is a deuterium-labeled analogue of olaparib used as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS analyses. It retains the inhibitory profile against PARP1 and PARP2 while providing a mass-shifted isotopic label for accurate quantitation in analytical workflows.

  • High isotopic labeling for reliable mass-based discrimination.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Potent PARP1/2 inhibitory profile (IC50s: PARP1 5 nM, PARP2 1 nM).
  • High purity for analytical accuracy (≈99.6%).
  • Soluble in common solvents such as DMSO and DMF.
  • Stable when stored under recommended low-temperature conditions.

Catalog No. 50-002-47783 Supplier Medchemexpress LLC Supplier No. HY10162S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olaparib-d8 is a deuterium-labeled analogue of olaparib used as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS analyses. It retains the inhibitory profile against PARP1 and PARP2 while providing a mass-shifted isotopic label for accurate quantitation in analytical workflows.

  • High isotopic labeling for reliable mass-based discrimination.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Potent PARP1/2 inhibitory profile (IC50s: PARP1 5 nM, PARP2 1 nM).
  • High purity for analytical accuracy (≈99.6%).
  • Soluble in common solvents such as DMSO and DMF.
  • Stable when stored under recommended low-temperature conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.