Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olaparib-d8 | 00-00-0 | 99.6% | 442.51 g·mol⁻¹ | C24H15D8FN4O3 | 5 MG
SDP

Catalog No. 5000248689
Encompass_Preferred

Olaparib-d8 is the deuterium-labeled analogue of the PARP inhibitor olaparib, supplied as a stable isotope internal standard for quantitative bioanalytical assays. It is intended for use in LC-MS/MS and other mass spectrometry workflows to improve accuracy and precision when measuring olaparib in biological matrices.

  • Deuterium-labeled internal standard for LC-MS/MS and quantitative assays.
  • High purity (99.6% by RP-HPLC/LCMS).
  • Provides an 8 Da mass shift for clear analyte/internal standard separation in mass spectrometry.
  • Suitable for pharmacokinetic, bioanalysis, and method validation studies.
  • Molecular weight 442.51 g·mol⁻¹ and formula C24H15D8FN4O3.
  • Supplied as a small-quantity analytical reference material for laboratory use.

Catalog No. 50-002-48689 Supplier Medchemexpress LLC Supplier No. HY10162S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olaparib-d8 is the deuterium-labeled analogue of the PARP inhibitor olaparib, supplied as a stable isotope internal standard for quantitative bioanalytical assays. It is intended for use in LC-MS/MS and other mass spectrometry workflows to improve accuracy and precision when measuring olaparib in biological matrices.

  • Deuterium-labeled internal standard for LC-MS/MS and quantitative assays.
  • High purity (99.6% by RP-HPLC/LCMS).
  • Provides an 8 Da mass shift for clear analyte/internal standard separation in mass spectrometry.
  • Suitable for pharmacokinetic, bioanalysis, and method validation studies.
  • Molecular weight 442.51 g·mol⁻¹ and formula C24H15D8FN4O3.
  • Supplied as a small-quantity analytical reference material for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.