Learn More
Medchemexpress LLC Olaquindox | 23696-28-8 | MFCD00210341 | 99.9% | 263.25 | C12H13N3O4 | 10 MG
SDP
Catalog No. 5000296405
Encompass_Preferred
Olaquindox is a quinoxaline-derivative antibiotic supplied as a research-grade solid. It is orally active and has been used historically as a growth promoter in swine; provided at high purity for laboratory and analytical applications.
  • High purity (99.9%) suitable for analytical use.
  • Orally active quinoxaline derivative with documented biological effects.
  • Solid, light yellow to green-yellow appearance for easy handling.
  • Suitable for research and analytical applications.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-96405 Supplier Medchemexpress LLC Supplier No. HYN046510MG
Olaquindox is a quinoxaline-derivative antibiotic supplied as a research-grade solid. It is orally active and has been used historically as a growth promoter in swine; provided at high purity for laboratory and analytical applications.
  • High purity (99.9%) suitable for analytical use.
  • Orally active quinoxaline derivative with documented biological effects.
  • Solid, light yellow to green-yellow appearance for easy handling.
  • Suitable for research and analytical applications.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.