Learn More
Medchemexpress LLC HY-10095 2mg , Olcegepant CAS:204697-65-4 Purity:>98%
SDP
Catalog No. 502259875
Encompass_Preferred
HY-10095 2mg Olcegepant CAS: 204697-65-4 Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9875 Supplier Medchemexpress LLC Supplier No. HY100952MG
HY-10095 2mg Olcegepant CAS: 204697-65-4 Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.