Learn More
Medchemexpress LLC Olcegepant hydrochloride | 586368-06-1 | 906.11 | C38H47Br2N9O5·HCl | 1 ML
SDP
Catalog No. 5000211270
Encompass_Preferred
Olcegepant hydrochloride is the hydrochloride salt of olcegepant, a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor used in neuroscience and pharmacology research. It is supplied in ready-to-use solution and solid+solvent formats for flexible laboratory use and includes analytical documentation for verification.
Catalog No. 50-002-11270 Supplier Medchemexpress LLC Supplier No. HY10095A10MM 1ML
Olcegepant hydrochloride is the hydrochloride salt of olcegepant, a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor used in neuroscience and pharmacology research. It is supplied in ready-to-use solution and solid+solvent formats for flexible laboratory use and includes analytical documentation for verification.