Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olcegepant hydrochloride | 586368-06-1 | MFCD07772306 | 99.1% | 906.11 | C38H47Br2N9O5·HCl | 50 MG
SDP

Catalog No. 5000211951
Encompass_Preferred

Olcegepant hydrochloride is a potent, selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor, provided as the hydrochloride salt for preclinical research use. The compound is supplied with reported high purity and is offered in multiple quantities and solution formats for laboratory studies.

  • Potent, selective antagonist of the CGRP1 receptor.
  • Reported high purity (~99.1%).
  • Available in multiple sizes, including 5 mg-500 mg, and as DMSO solutions.
  • Molecular formula C38H47Br2N9O5·HCl; molecular weight 906.11 g/mol.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-11951 Supplier Medchemexpress LLC Supplier No. HY10095A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olcegepant hydrochloride is a potent, selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor, provided as the hydrochloride salt for preclinical research use. The compound is supplied with reported high purity and is offered in multiple quantities and solution formats for laboratory studies.

  • Potent, selective antagonist of the CGRP1 receptor.
  • Reported high purity (~99.1%).
  • Available in multiple sizes, including 5 mg-500 mg, and as DMSO solutions.
  • Molecular formula C38H47Br2N9O5·HCl; molecular weight 906.11 g/mol.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.