Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olcegepant hydrochloride | 586368-06-1 | MFCD30738010 | 99.1% | 906.11 | C38H47Br2N9O5·HCl | 10 MG
SDP

Catalog No. 5000211266
Encompass_Preferred

Olcegepant hydrochloride is a selective non-peptide antagonist of the calcitonin gene-related peptide (CGRP) receptor supplied as the hydrochloride salt for laboratory research. It is provided as a high-purity solid for use in neuroscience and GPCR pharmacology studies.

  • Hydrochloride salt form, suitable for aqueous formulations.
  • CAS number 586368-06-1.
  • Molecular formula C38H47Br2N9O5·HCl and molecular weight 906.11.
  • High purity (~99.1%) suitable for analytical and preparative studies.
  • Available in small-scale quantities, e.g., 10 MG, for laboratory use.
  • Used to probe calcitonin gene-related peptide (CGRP) receptor signaling in preclinical research.

Catalog No. 50-002-11266 Supplier Medchemexpress LLC Supplier No. HY10095A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olcegepant hydrochloride is a selective non-peptide antagonist of the calcitonin gene-related peptide (CGRP) receptor supplied as the hydrochloride salt for laboratory research. It is provided as a high-purity solid for use in neuroscience and GPCR pharmacology studies.

  • Hydrochloride salt form, suitable for aqueous formulations.
  • CAS number 586368-06-1.
  • Molecular formula C38H47Br2N9O5·HCl and molecular weight 906.11.
  • High purity (~99.1%) suitable for analytical and preparative studies.
  • Available in small-scale quantities, e.g., 10 MG, for laboratory use.
  • Used to probe calcitonin gene-related peptide (CGRP) receptor signaling in preclinical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.