Learn More
Medchemexpress LLC Olcegepant hydrochloride | 586368-06-1 | MFCD30738010 | 99.3% | 906.11 g/mol | C38H47Br2N9O5.HCl | 100 MG
SDP
Catalog No. 5000212228
Encompass_Preferred
Olcegepant hydrochloride is a selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor used in neuroscience and pharmacology research. Supplied as a stable solid with high reported purity, it is employed in assays to investigate CGRP signaling, receptor pharmacology, and migraine-related mechanisms.
  • Selective CGRP1 receptor antagonist suitable for receptor studies.
  • High purity appropriate for research applications.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available in multiple pack sizes for experimental flexibility.
  • Provided as a stable solid powder for convenient storage and handling.
Catalog No. 50-002-12228 Supplier Medchemexpress LLC Supplier No. HY10095A100MG
Olcegepant hydrochloride is a selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor used in neuroscience and pharmacology research. Supplied as a stable solid with high reported purity, it is employed in assays to investigate CGRP signaling, receptor pharmacology, and migraine-related mechanisms.
  • Selective CGRP1 receptor antagonist suitable for receptor studies.
  • High purity appropriate for research applications.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available in multiple pack sizes for experimental flexibility.
  • Provided as a stable solid powder for convenient storage and handling.