Learn More
Apexbio Technology LLC Oleamide 301-02-0 50mg
SDP
Catalog No. 502240314
Encompass_Preferred
Oleamide (CAS 301-02-0) is an endogenous fatty acid amide synthesized de novo within the mammalian nervous system and detected in human plasma It modulates neuronal signaling by inhibiting specific signal transduction pathways and altering the activity of certain enzymes involved in neurophysiological regulation Oleamide has been extensively utilized in research models investigating neurodegenerative conditions and sleep disorders where its actions provide insights into the modulation of neural circuitry and associated biochemical processes
This item is not returnable. View return policy
Catalog No. 50-224-0314 Supplier Apexbio Technology LLC Supplier No. B648450
Oleamide (CAS 301-02-0) is an endogenous fatty acid amide synthesized de novo within the mammalian nervous system and detected in human plasma It modulates neuronal signaling by inhibiting specific signal transduction pathways and altering the activity of certain enzymes involved in neurophysiological regulation Oleamide has been extensively utilized in research models investigating neurodegenerative conditions and sleep disorders where its actions provide insights into the modulation of neural circuitry and associated biochemical processes