Learn More
Medchemexpress LLC Olean-12-en-29-oic acid, 3-hydroxy-11-oxo-, (3β,20β)- | 471-53-4 | 99.9% | 470.68 | 10 G
SDP
Catalog No. 5000427885
Encompass_Preferred
18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhiza uralensis and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.
  • Anti-ulcerative
  • Anti-inflammatory
  • Antiproliferative properties
Catalog No. 50-004-27885 Supplier Medchemexpress LLC Supplier No. HYN018010G
18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhiza uralensis and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.
  • Anti-ulcerative
  • Anti-inflammatory
  • Antiproliferative properties