Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olean-12-en-3-ol, 3-acetate, (3β)- | 1616-93-9 | MFCD00200955 | 99.9% | 468.75 g/mol | C32H52O2 | 10 MG
SDP

Catalog No. 5000289492
Encompass_Preferred

β-Amyrin acetate is a pentacyclic triterpenoid research reagent reported to have anti-inflammatory, antifungal, anti-diabetic, and anti-hyperlipidemic activities. It is used as a purified standard for in vitro and in vivo pharmacology and analytical studies.

  • Molecular formula C32H52O2, molecular weight 468.75 g/mol.
  • CAS number 1616-93-9.
  • Purity 99.89%.
  • Reported biological activities: anti-inflammatory, antifungal, anti-diabetic, anti-hyperlipidemic.
  • Available in small research pack sizes (5 mg, 10 mg, 50 mg, 100 mg).
  • Suitable as an analytical standard or pharmacology research reagent.

Catalog No. 50-002-89492 Supplier Medchemexpress LLC Supplier No. HYN292310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Amyrin acetate is a pentacyclic triterpenoid research reagent reported to have anti-inflammatory, antifungal, anti-diabetic, and anti-hyperlipidemic activities. It is used as a purified standard for in vitro and in vivo pharmacology and analytical studies.

  • Molecular formula C32H52O2, molecular weight 468.75 g/mol.
  • CAS number 1616-93-9.
  • Purity 99.89%.
  • Reported biological activities: anti-inflammatory, antifungal, anti-diabetic, anti-hyperlipidemic.
  • Available in small research pack sizes (5 mg, 10 mg, 50 mg, 100 mg).
  • Suitable as an analytical standard or pharmacology research reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.