Learn More
Medchemexpress LLC HY-116010 10mg Medchemexpress, Oleandomycin CAS:3922-90-5 Purity:>98%
SDP
Catalog No. 501969402
Encompass_Preferred
Medchemexpress, HY-116010 10mg Oleandomycin CAS:3922-90-5 Oleandomycin is a macrolide antibiotic structurally closely related to Erythromycin. Oleandomycin is similar to Erythromycin with antimicrobial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9402 Supplier Medchemexpress LLC Supplier No. HY11601010MG
Medchemexpress, HY-116010 10mg Oleandomycin CAS:3922-90-5 Oleandomycin is a macrolide antibiotic structurally closely related to Erythromycin. Oleandomycin is similar to Erythromycin with antimicrobial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.