Learn More
Selleck Chemical LLC Oleandrin
SDP
Catalog No. 5000000974
Encompass_Preferred
Oleandrin (PBI-05204 Foliandrin Neriolin Neriostene Folinerin Corrigen Neriol) is an inhibitor of Na /K -ATPase with IC50 of 0 62 M Oleandrin induces apoptosis and reduces migration of human glioma cells in vitro
Catalog No. 50-000-00974 Supplier Selleck Chemical LLC Supplier No. S69185MG
Oleandrin (PBI-05204 Foliandrin Neriolin Neriostene Folinerin Corrigen Neriol) is an inhibitor of Na /K -ATPase with IC50 of 0 62 M Oleandrin induces apoptosis and reduces migration of human glioma cells in vitro