Learn More
TARGETMOL CHEMICALS INC OLEANDRIN 5MG
SDP
Catalog No. 502579717
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Oleandrin (Folinerin) the principal cardiac glycoside component of PBI-524 can quantitatively account for regulation of BDNF at both the protein and transcriptional levels. 2. Oleandrin are known to inhibit the Na K-ATPase pump resulting in a consequent increase in calcium influx in heart muscle. 3. Oleandrin has stronger anti-proliferative activity in undifferentiated CaCO-2 cells (IC50 8.25 nM) causes an autophagic cell death and altered ERK phosphorylation in undifferentiated. purity: 99%
Catalog No. 50-257-9717 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S08905MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Oleandrin (Folinerin) the principal cardiac glycoside component of PBI-524 can quantitatively account for regulation of BDNF at both the protein and transcriptional levels. 2. Oleandrin are known to inhibit the Na K-ATPase pump resulting in a consequent increase in calcium influx in heart muscle. 3. Oleandrin has stronger anti-proliferative activity in undifferentiated CaCO-2 cells (IC50 8.25 nM) causes an autophagic cell death and altered ERK phosphorylation in undifferentiated. purity: 99%