Learn More
Apexbio Technology LLC Oleanolic acid(Synonyms: Oleanic acid, Caryophyllin, Astrantiagenin, Giganteumgenin, Caryophyllinic acid, 3β-Hydroxyolean-12-en-28-oic acid), 100mg, CAS: 508-02-1.
SDP
Catalog No. 5000061143
Encompass_Preferred
Oleanolic acid (CAS 508-02-1) is a naturally occurring triterpenoid isolated primarily from garlic and Phytolacca americana It exerts anti-HIV activity through induction of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) enzymes involved in inflammatory and immune responses Due to its mechanism of modulating these targets oleanolic acid serves as a valuable compound in biomedical research aimed at exploring novel antiviral strategies and immune pathway modulation
Catalog No. 50-000-61143 Supplier Apexbio Technology LLC Supplier No. N1826100
Oleanolic acid (CAS 508-02-1) is a naturally occurring triterpenoid isolated primarily from garlic and Phytolacca americana It exerts anti-HIV activity through induction of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) enzymes involved in inflammatory and immune responses Due to its mechanism of modulating these targets oleanolic acid serves as a valuable compound in biomedical research aimed at exploring novel antiviral strategies and immune pathway modulation