Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleanolic Acid, 98.4% (HPLC), MP Biomedicals

Catalog No. ICN15398180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15398180 100 mg
ICN15398190 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15398180 Supplier MP Biomedicals, Inc Supplier No. 0215398180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 508-02-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.711
InChI Key MIJYXULNPSFWEK-GTOFXWBISA-N
Synonym oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
PubChem CID 10494
ChEBI CHEBI:37659
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C

Specifications

Color White
Assay Percent Range 0.984
Quantity 100 mg
Formula Weight 456.7
Physical Form Powder
Chemical Name or Material Oleanolic Acid, For HPLC analysis
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.