Learn More
Selleck Chemical LLC Oleanolic Acid 100mg 508-02-1 Caryophyllin
SDP
Supplier:  Selleck Chemical LLC S2334100MG
Encompass_Preferred
Oleanolic Acid (Caryophyllin) is a non-toxic, hepatoprotective triterpenoid found in Phytolacca Americana, which exerts antitumor and antiviral properties. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-543-0