Learn More
Medchemexpress LLC Oleanolic acid hemiphthalate disodium salt | 168770-31-8 | 98.1% | 1 ML
SDP
Catalog No. 5000339737
Encompass_Preferred
Oleanolic acid hemiphthalate disodium salt is a hemiphthalate derivative of Oleanolic acid, exhibiting anti-inflammatory and potential anti-ulcer activities. This product is strictly for research purposes and not for human or animal medical, clinical diagnostic, or food applications.
  • Stable under recommended storage conditions.
  • Soluble in DMSO, requiring sonication.
  • Shows anti-inflammatory properties.
  • Potential for anti-ulcer activities.
  • Store solid product at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for up to 6 months, or -20°C for up to 1 month.
Catalog No. 50-003-39737 Supplier Medchemexpress LLC Supplier No. HY12869510MM/1ML
Oleanolic acid hemiphthalate disodium salt is a hemiphthalate derivative of Oleanolic acid, exhibiting anti-inflammatory and potential anti-ulcer activities. This product is strictly for research purposes and not for human or animal medical, clinical diagnostic, or food applications.
  • Stable under recommended storage conditions.
  • Soluble in DMSO, requiring sonication.
  • Shows anti-inflammatory properties.
  • Potential for anti-ulcer activities.
  • Store solid product at 4°C, sealed, away from moisture.
  • Store in solvent at -80°C for up to 6 months, or -20°C for up to 1 month.