Learn More
Medchemexpress LLC Oleanolic aldehyde | 17020-22-3 | 440.70 | 1 MG
SDP
Catalog No. 5000414582
Encompass_Preferred
Oleanolic aldehyde is an antimicrobial compound effective against oral bacteria like *Streptococcus mutans* and *Porphyromonas gingivalis*, which are implicated in dental caries and periodontal disease. This compound, classified as a triterpene, is naturally derived from plants such as *Vitis vinifera* cv. Zalema.
  • Antimicrobial properties
  • Inhibits *Streptococcus mutans* (MIC 488 μg/mL)
  • Inhibits *Porphyromonas gingivalis* (MIC 250 μg/mL)
  • Demonstrates growth inhibition in B16 2F2 epithelial cells
  • Exhibits antiproliferative activity against +SA mammary epithelial cells
  • Plant-derived triterpene
Catalog No. 50-004-14582 Supplier Medchemexpress LLC Supplier No. HYN122401MG
Oleanolic aldehyde is an antimicrobial compound effective against oral bacteria like *Streptococcus mutans* and *Porphyromonas gingivalis*, which are implicated in dental caries and periodontal disease. This compound, classified as a triterpene, is naturally derived from plants such as *Vitis vinifera* cv. Zalema.
  • Antimicrobial properties
  • Inhibits *Streptococcus mutans* (MIC 488 μg/mL)
  • Inhibits *Porphyromonas gingivalis* (MIC 250 μg/mL)
  • Demonstrates growth inhibition in B16 2F2 epithelial cells
  • Exhibits antiproliferative activity against +SA mammary epithelial cells
  • Plant-derived triterpene