Learn More
Medchemexpress LLC Oleanolic aldehyde | 17020-22-3 | 440.70 | 5 MG
SDP
Catalog No. 5000415035
Encompass_Preferred
Oleanolic aldehyde is an antimicrobial compound that inhibits oral bacteria. It is specifically effective against *Streptococcus mutans* and *Porphyromonas gingivalis*, which are associated with dental caries and periodontal disease.
  • Antimicrobial compound
  • Inhibits oral bacteria such as *Streptococcus mutans* and *Porphyromonas gingivalis*
  • Associated with dental caries and periodontal disease research
Catalog No. 50-004-15035 Supplier Medchemexpress LLC Supplier No. HYN122405MG
Oleanolic aldehyde is an antimicrobial compound that inhibits oral bacteria. It is specifically effective against *Streptococcus mutans* and *Porphyromonas gingivalis*, which are associated with dental caries and periodontal disease.
  • Antimicrobial compound
  • Inhibits oral bacteria such as *Streptococcus mutans* and *Porphyromonas gingivalis*
  • Associated with dental caries and periodontal disease research