Learn More
Selleck Chemical LLC Oleic Acid-100mg
SDP
Catalog No. 502390037
Encompass_Preferred
Oleic acid (Oleate, 9-Octadecenoic Acid, Elaidoic acid) is a fatty acid that occurs naturally in various animal and vegetable fats and oils. It is used commercially in the preparation of oleates and lotions, and as a pharmaceutical solvent. Oleic acid is an abundant monounsaturated fatty acid. Oleic acid is a Na+/K+ ATPase activator.
Catalog No. 50-239-0037 Supplier Selleck Chemical LLC Supplier No. S4707100MG
Oleic acid (Oleate, 9-Octadecenoic Acid, Elaidoic acid) is a fatty acid that occurs naturally in various animal and vegetable fats and oils. It is used commercially in the preparation of oleates and lotions, and as a pharmaceutical solvent. Oleic acid is an abundant monounsaturated fatty acid. Oleic acid is a Na+/K+ ATPase activator.