Learn More
Medchemexpress LLC Oleic acid | 112-80-1 | 282.46 | 5 MG
SDP
Catalog No. 5000415926
Encompass_Preferred
Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid in human adipocytes and other tissues. Oleic acid promotes cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation. It also inhibits cancer cell growth and survival in low metastatic carcinoma cells like gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines. This product is intended for research and analytical applications.
  • Abundant monounsaturated fatty acid
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration in high metastatic cancer cells
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells
  • Intended for research and analytical applications
Catalog No. 50-004-15926 Supplier Medchemexpress LLC Supplier No. HYN14465MG
Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid in human adipocytes and other tissues. Oleic acid promotes cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation. It also inhibits cancer cell growth and survival in low metastatic carcinoma cells like gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines. This product is intended for research and analytical applications.
  • Abundant monounsaturated fatty acid
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration in high metastatic cancer cells
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells
  • Intended for research and analytical applications