Learn More
Selleck Chemical LLC Oleic Acid S4707-1g
SDP
Catalog No. 503109414
Encompass_Preferred
Oleic acid (Oleate 9-Octadecenoic Acid Elaidoic acid) is a fatty acid that occurs naturally in various animal and vegetable fats and oils It is used commercially in the preparation of oleates and lotions and as a pharmaceutical solvent Oleic acid is an abundant monounsaturated fatty acid Oleic acid is a Na /K ATPase activator Oleic Acid can be used to induce animal models of Renal Failure
Catalog No. 50-310-9414 Supplier Selleck Chemical LLC Supplier No. S47071G
Oleic acid (Oleate 9-Octadecenoic Acid Elaidoic acid) is a fatty acid that occurs naturally in various animal and vegetable fats and oils It is used commercially in the preparation of oleates and lotions and as a pharmaceutical solvent Oleic acid is an abundant monounsaturated fatty acid Oleic acid is a Na /K ATPase activator Oleic Acid can be used to induce animal models of Renal Failure