Learn More
Cayman Chemical Oleic Acid-13C, Purity: ≥98%, CAS: 82005-44-5, 1 mg
SDP
Catalog No. 502938736
Encompass_Preferred
An internal standard for the quantification of oleic acid by GC- or LC-MS
Catalog No. 50-293-8736 Supplier Cayman Chemical Supplier No. 278691 MG
An internal standard for the quantification of oleic acid by GC- or LC-MS