Learn More
Medchemexpress LLC Oleic acid-13C | 82005-44-5 | 283.45 | 5 MG
SDP
Catalog No. 5000414861
Encompass_Preferred
Oleic acid-13C is the 13C labeled Oleic acid (HY-N1446). Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid and acts as a Na+/K+ ATPase activator.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-14861 Supplier Medchemexpress LLC Supplier No. HYN1446S5MG
Oleic acid-13C is the 13C labeled Oleic acid (HY-N1446). Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid and acts as a Na+/K+ ATPase activator.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.