Learn More
Medchemexpress LLC Oleic acid-13C18 | 287100-82-7 | 300.33 | 25 MG
SDP
Catalog No. 5000389484
Encompass_Preferred
Oleic acid-13C18 is the 13C labeled Oleic acid, an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Abundant monounsaturated fatty acid
  • Na+/K+ ATPase activator
Catalog No. 50-003-89484 Supplier Medchemexpress LLC Supplier No. 25MGHYN1446S2
Oleic acid-13C18 is the 13C labeled Oleic acid, an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Abundant monounsaturated fatty acid
  • Na+/K+ ATPase activator