Learn More
Medchemexpress LLC Oleic acid-13C18 | 287100-82-7 | 300.33 | 5 MG
SDP
Catalog No. 5000415361
Encompass_Preferred
Oleic acid-13C18 is the 13C labeled version of Oleic acid (HY-N1446). Oleic acid (9-cis-Octadecenoic acid) is a common monounsaturated fatty acid and acts as a Na+/K+ ATPase activator.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.9%.
Catalog No. 50-004-15361 Supplier Medchemexpress LLC Supplier No. HYN1446S25MG
Oleic acid-13C18 is the 13C labeled version of Oleic acid (HY-N1446). Oleic acid (9-cis-Octadecenoic acid) is a common monounsaturated fatty acid and acts as a Na+/K+ ATPase activator.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.9%.