Learn More
Sigma Aldrich Fine Chemicals Biosciences Oleic acid 13C18 99 atom1G
Catalog No. 502902869
Encompass
Oleic acid-13C18 is a 13C labeled fatty acid. The disposition of 13C18 -oleic acid following in vivo intravenous and oral administration has been determined by liquid chromatography-mass spectrometry.
Catalog No. 50-290-2869 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4904311G
Add to Cart
Add to Cart
Request a Quote
Oleic acid-13C18 is a 13C labeled fatty acid. The disposition of 13C18 -oleic acid following in vivo intravenous and oral administration has been determined by liquid chromatography-mass spectrometry.