Learn More
Cayman Chemical OleIc AcId-13C5 25mg
SDP
Catalog No. 502892844
Encompass_Preferred
An internal standard for the quantification of oleic acid by GC- or LC-MS
Catalog No. 50-289-2844 Supplier Cayman Chemical Supplier No. 900408925 MG
An internal standard for the quantification of oleic acid by GC- or LC-MS