Learn More
Cayman Chemical Oleic Acid-13C5, Purity: ≥95%, CAS: 1255644-48-4, 5 mg
SDP
Catalog No. 502895921
Encompass_Preferred
An internal standard for the quantification of oleic acid by GC- or LC-MS
Catalog No. 50-289-5921 Supplier Cayman Chemical Supplier No. 90040895 MG
An internal standard for the quantification of oleic acid by GC- or LC-MS