Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oleic Acid

Catalog No. 277281LR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-728-1LR 1 L
27-728-25LR 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-728-1LR Supplier Sigma Aldrich Supplier No. 277281LR
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oleic Acid

Specifications

CAS 112-80-1
Melting Point 13°C to 14°C
Density 0.89 g/mL at 25°C
Boiling Point 194°C to 195°C (1.2 mmHg, lit.)
Molecular Formula C18H34O2
Refractive Index n20/D 1.459
Linear Formula CH3(CH2)7CH=CH(CH2)7COOH
Quantity 1 L
MDL Number MFCD00064242
Synonym cis-9-Octadecenoic acid; Elainic acid
Molecular Weight (g/mol) 282.46 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.