Learn More
Sigma Aldrich Fine Chemicals Biosciences Oleic acid, 112-80-1, MFCD00064242, 5 g
Catalog No. NC0842203
Encompass
Linear Formula: CH3(CH2)7CH=CH(CH2)7COOH, Molecular Weight: 282.46, ≥99% (GC), bp: 194-195 °C, mp: 13-14 °C, Synonym: cis-9-Octadecenoic acid, Elainic acid.
Catalog No. NC0842203 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. O10085G
Linear Formula: CH3(CH2)7CH=CH(CH2)7COOH, Molecular Weight: 282.46, ≥99% (GC), bp: 194-195 °C, mp: 13-14 °C, Synonym: cis-9-Octadecenoic acid, Elainic acid.