Learn More
Cayman Chemical Oleic Acid, 112-80-1, 500 mg
SDP
Supplier:  Cayman Chemical 90260500
Encompass_Preferred
A monounsaturated fatty acid; a major component of membrane phospholipids; inhibits collagen-stimulated platelet aggregation at 10 µg/ml; inhibits fMLF-induced neutrophil aggregation and degranulation at 5 µM; induces release of intracellular calcium in human platelets at 60 µM; increases TNF-α, IL-8, IL-6, and IL-1β production, neutrophil accumulation, and apoptotic and necrotic cell death in mouse lung
Catalog No. NC0035766