Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic acid, 71%, MP Biomedicals™

Catalog No. ICN10250090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10250090 500 g
ICN10250091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10250090 Supplier MP Biomedicals, Inc Supplier No. 0210250090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Color Colorless to White
Density 0.895g/mL
Assay Percent Range 0.71
Quantity 500 g
Formula Weight 282.52
Physical Form Liquid To Solid
Chemical Name or Material Oleic acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.