Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleic acid (9-cis-Octadecenoic acid) | 112-80-1 | 282.46 | 1 ML
SDP

Catalog No. 5000415487
Encompass_Preferred

Oleic acid is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator.

  • High purity of 99.97%.
  • Appears as a solid below 13°C and a liquid above 14°C.
  • Colorless to light yellow.
  • Soluble in ethanol, 0.1 M NaOH, and DMSO.
  • Classified as ketones, aldehydes, and acids.
  • Derived from an endogenous metabolite.
  • Ships at room temperature in the continental US.
  • Storage conditions vary based on form: -20°C for 3 years (pure) or -80°C for 6 months (in solvent).

Catalog No. 50-004-15487 Supplier Medchemexpress LLC Supplier No. HYN144610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleic acid is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator.

  • High purity of 99.97%.
  • Appears as a solid below 13°C and a liquid above 14°C.
  • Colorless to light yellow.
  • Soluble in ethanol, 0.1 M NaOH, and DMSO.
  • Classified as ketones, aldehydes, and acids.
  • Derived from an endogenous metabolite.
  • Ships at room temperature in the continental US.
  • Storage conditions vary based on form: -20°C for 3 years (pure) or -80°C for 6 months (in solvent).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.