Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012507 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2507 5 mL
11-101-2506 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2507 Supplier MilliporeSigma Supelco Supplier No. 750905ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleic acid is found to be an important component of olive oil, which can help in the regulation of many biological processes. It acts as a penetration enhancer by increasing lipid fluidity thereby increasing the skin permeability.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 112-80-1
Vapor Pressure 1 mmHg (176°C)
Density 0.89 g/mL (at 25°C)
Shipping Condition Dry ice
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOH
MDL Number MFCD00064242
Melting Point 13°C to 14°C
Boiling Point 194°C to 195°C (1.2 mmHg)
Synonym cis-9-Octadecenoic acid; Elainic acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 282.46
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.