Learn More
Cayman Chemical OleIc AcId Alkyn 100UG
SDP
Catalog No. 502893322
Encompass_Preferred
A form of oleic acid with an ω-terminal alkyne for use in click chemistry reactions
Catalog No. 50-289-3322 Supplier Cayman Chemical Supplier No. 9002078100 UG
A form of oleic acid with an ω-terminal alkyne for use in click chemistry reactions