Learn More
Medchemexpress LLC Oleic acid alkyne | 151333-45-8 | 100.0% | 10 MG
SDP
Catalog No. 5000342424
Encompass_Preferred
Oleic acid alkyne is a form of oleic acid that incorporates an acetylene group. This terminal alkyne group allows for its use in click chemical ligation reactions. Oleic acid itself can undergo hydroxylation through a microsomal cytochrome P-450-dependent system (ω-OAH). The incorporation of fluorescent or biotin labels into oleic acid via click chemistry reactions enables the analysis of its metabolism and biological activity.
  • The terminal alkyne group facilitates click chemical ligation reactions.
  • It can be used to introduce fluorescent or biotin labels for studying oleic acid metabolism and biological activity.
  • For research use only.
Catalog No. 50-003-42424 Supplier Medchemexpress LLC Supplier No. HY12711210MG
Oleic acid alkyne is a form of oleic acid that incorporates an acetylene group. This terminal alkyne group allows for its use in click chemical ligation reactions. Oleic acid itself can undergo hydroxylation through a microsomal cytochrome P-450-dependent system (ω-OAH). The incorporation of fluorescent or biotin labels into oleic acid via click chemistry reactions enables the analysis of its metabolism and biological activity.
  • The terminal alkyne group facilitates click chemical ligation reactions.
  • It can be used to introduce fluorescent or biotin labels for studying oleic acid metabolism and biological activity.
  • For research use only.