Learn More
Medchemexpress LLC Oleic acid alkyne | 151333-45-8 | 99.8% | 500 UG
SDP
Catalog No. 5000342422
Encompass_Preferred
Oleic acid alkyne is oleic acid with an acetylene group. It is designed for use in click chemical ligation reactions.
  • Terminal alkyne group for click chemical ligation reactions
  • Can be hydroxylated by a microsomal cytochrome P-450-dependent system (ω-OAH)
  • Enables introduction of fluorescent or biotin-labeled oleic acid for metabolic analysis
  • Facilitates the study of oleic acid metabolism and biological activity
Catalog No. 50-003-42422 Supplier Medchemexpress LLC Supplier No. HY127112500UG
Oleic acid alkyne is oleic acid with an acetylene group. It is designed for use in click chemical ligation reactions.
  • Terminal alkyne group for click chemical ligation reactions
  • Can be hydroxylated by a microsomal cytochrome P-450-dependent system (ω-OAH)
  • Enables introduction of fluorescent or biotin-labeled oleic acid for metabolic analysis
  • Facilitates the study of oleic acid metabolism and biological activity