Learn More
Medchemexpress LLC Oleic acid (cis-9-octadecenoic acid) | 112-80-1 | MFCD00064242 | 100.0% | 10 MG
SDP
Catalog No. 5000296158
Encompass_Preferred
Oleic acid (cis-9-octadecenoic acid) is an abundant monounsaturated fatty acid commonly used in biochemical and cell biology research. It is reported to activate Na+/K+ ATPase and is used to study membrane dynamics, lipid metabolism, and cell signaling. The compound is characterized by CAS 112-80-1, molecular formula C18H34O2, and molecular weight 282.46 g/mol; follow recommended storage for pure material and solvent solutions.
  • Monounsaturated fatty acid (cis-9-octadecenoic acid).
  • Reported Na+/K+ ATPase activator.
  • Suitable for studies of membrane biology, lipid metabolism, and cell signaling.
  • Documented storage recommendations for pure material and in solvent.
  • Characterized by CAS 112-80-1 and molecular weight 282.46 g/mol.
Catalog No. 50-002-96158 Supplier Medchemexpress LLC Supplier No. HYN144610MG
Oleic acid (cis-9-octadecenoic acid) is an abundant monounsaturated fatty acid commonly used in biochemical and cell biology research. It is reported to activate Na+/K+ ATPase and is used to study membrane dynamics, lipid metabolism, and cell signaling. The compound is characterized by CAS 112-80-1, molecular formula C18H34O2, and molecular weight 282.46 g/mol; follow recommended storage for pure material and solvent solutions.
  • Monounsaturated fatty acid (cis-9-octadecenoic acid).
  • Reported Na+/K+ ATPase activator.
  • Suitable for studies of membrane biology, lipid metabolism, and cell signaling.
  • Documented storage recommendations for pure material and in solvent.
  • Characterized by CAS 112-80-1 and molecular weight 282.46 g/mol.