Learn More
*IMAGION BIOSYSTEMS INC PrecisionMRX Oleic Acid Coated 25nm Iron Oxide Nanoparticles – 50mg
Catalog No. NC1837699
Encompass
Superparamagnetic iron oxide nanoparticles with a diameter of 24 – 26 nm and a low size dispersity (≤7%) as measured by Small Angle X-ray Scattering (SAXS). The cores have an oleic acid coating and are easily dispersed in non-polar solvents such as hexane, toluene, chloroform, dichloromethane, etc.
Catalog No. NC1837699 Supplier *IMAGION BIOSYSTEMS INC Supplier No. 0A050S
Superparamagnetic iron oxide nanoparticles with a diameter of 24 – 26 nm and a low size dispersity (≤7%) as measured by Small Angle X-ray Scattering (SAXS). The cores have an oleic acid coating and are easily dispersed in non-polar solvents such as hexane, toluene, chloroform, dichloromethane, etc.