Learn More
Medchemexpress LLC Oleic acid-d17 | 223487-44-3 | 299.57 | 500 UG
SDP
Catalog No. 5000415710
Encompass_Preferred
Oleic acid-d17 is the deuterium labeled Oleic acid, an abundant monounsaturated fatty acid that acts as a Na+/K+ ATPase activator. This product is suitable for various analytical techniques.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-15710 Supplier Medchemexpress LLC Supplier No. HYN1446S3500UG
Oleic acid-d17 is the deuterium labeled Oleic acid, an abundant monounsaturated fatty acid that acts as a Na+/K+ ATPase activator. This product is suitable for various analytical techniques.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs