Learn More
Medchemexpress LLC Oleic acid-d2 | 5711-29-5 | 284.47 | 100 MG
SDP
Catalog No. 5000414665
Encompass_Preferred
Oleic acid-d2 is a deuterium labeled oleic acid, an abundant monounsaturated fatty acid, and a Na+/K+ ATPase activator. It can be utilized as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity (99.6% or 99.8%)
  • Appears as a colorless to light yellow liquid
Catalog No. 50-004-14665 Supplier Medchemexpress LLC Supplier No. HYN1446S1100MG
Oleic acid-d2 is a deuterium labeled oleic acid, an abundant monounsaturated fatty acid, and a Na+/K+ ATPase activator. It can be utilized as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity (99.6% or 99.8%)
  • Appears as a colorless to light yellow liquid