Learn More
Medchemexpress LLC Oleic acid-d2 (9-cis-Octadecenoic acid-d2) | 5711-29-5 | 284.47 | 5 MG
SDP
Catalog No. 5000414863
Encompass_Preferred
Oleic acid-d2 is the deuterium labeled Oleic acid (HY-N1446). It is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-14863 Supplier Medchemexpress LLC Supplier No. HYN1446S15MG
Oleic acid-d2 is the deuterium labeled Oleic acid (HY-N1446). It is an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. Stable heavy isotopes, such as deuterium, have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.