Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleic acid-d2 (9-cis-octadecenoic acid-d2) | 5711-29-5 | 99.6% | 10 MG
SDP

Catalog No. 5000296131
Encompass_Preferred

Oleic acid-d2 is the deuterium-labeled form of oleic acid (9-cis-octadecenoic acid) provided as a neat liquid for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is reported to modulate Na+/K+ ATPase activity and is supplied with high isotopic and analytical purity for research applications.

  • Deuterium-labeled internal standard for quantitative mass spectrometry and NMR.
  • High analytical purity (99.6%).
  • Molecular formula C18H32D2O2 and molecular weight 284.47.
  • Available in research-scale quantities, including 10 mg vials.
  • Colorless to light yellow liquid; density approximately 0.896 g/cm3.

Catalog No. 50-002-96131 Supplier Medchemexpress LLC Supplier No. HYN1446S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleic acid-d2 is the deuterium-labeled form of oleic acid (9-cis-octadecenoic acid) provided as a neat liquid for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is reported to modulate Na+/K+ ATPase activity and is supplied with high isotopic and analytical purity for research applications.

  • Deuterium-labeled internal standard for quantitative mass spectrometry and NMR.
  • High analytical purity (99.6%).
  • Molecular formula C18H32D2O2 and molecular weight 284.47.
  • Available in research-scale quantities, including 10 mg vials.
  • Colorless to light yellow liquid; density approximately 0.896 g/cm3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.