Learn More
Medchemexpress LLC Oleic acid-d2 (9-cis-Octadecenoic acid-d2; 9Z-Octadecenoic acid-d2) | 5711-29-5 | 284.47 | 1 MG
SDP
Catalog No. 5000414862
Encompass_Preferred
Oleic acid-d2 is the deuterium labeled version of Oleic acid, an abundant monounsaturated fatty acid. It acts as a Na+/K+ ATPase activator. This compound is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity (99.6%).
  • Stable heavy isotopes are incorporated for quantitation during drug development.
Catalog No. 50-004-14862 Supplier Medchemexpress LLC Supplier No. HYN1446S11MG
Add to Cart
Add to Cart
Oleic acid-d2 is the deuterium labeled version of Oleic acid, an abundant monounsaturated fatty acid. It acts as a Na+/K+ ATPase activator. This compound is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity (99.6%).
  • Stable heavy isotopes are incorporated for quantitation during drug development.