Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleic acid-d9 | 2687960-84-3 | MFCD00133433 | 95.0% | 291.52 g/mol | C18H25D9O2 | 10 MG
SDP

Catalog No. 5000296481
Encompass_Preferred

Oleic acid-d9 is a deuterium-labeled oleic acid used as a stable isotope standard and tracer in biochemical and analytical workflows. It is a monounsaturated fatty acid that acts as a Na+/K+ ATPase activator and is supplied as a colorless to light yellow liquid for analytical applications.

  • Deuterium-labeled for mass spectrometry and tracer studies.
  • Molecular formula C18H25D9O2; molecular weight 291.52 g/mol.
  • Typical purity 95.0%; batch-specific higher purities reported.
  • Available in 10 MG, 50 MG, 100 MG sizes suitable for analytical use.
  • Liquid, colorless to light yellow physical form.
  • Store at -20 °C for long-term; in solvent store at -80 °C for extended stability.

Catalog No. 50-002-96481 Supplier Medchemexpress LLC Supplier No. HYN1446S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleic acid-d9 is a deuterium-labeled oleic acid used as a stable isotope standard and tracer in biochemical and analytical workflows. It is a monounsaturated fatty acid that acts as a Na+/K+ ATPase activator and is supplied as a colorless to light yellow liquid for analytical applications.

  • Deuterium-labeled for mass spectrometry and tracer studies.
  • Molecular formula C18H25D9O2; molecular weight 291.52 g/mol.
  • Typical purity 95.0%; batch-specific higher purities reported.
  • Available in 10 MG, 50 MG, 100 MG sizes suitable for analytical use.
  • Liquid, colorless to light yellow physical form.
  • Store at -20 °C for long-term; in solvent store at -80 °C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.